missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Polyethylene Glycol 1500, NF, Spectrum™ Chemical
PO120, 25322-68-3, H(OCH2 CH2 )n OH
Chemical Identifiers
CAS
25322-68-3
Molecular Formula
(C2H4O)n
Molecular Weight (g/mol)
62.07
MDL Number
MFCD01779601
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name
ethane-1,2-diol
SMILES
[H]OCCO
View More Specs
Products
2
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
18605308
View Documents
Spectrum Chemical Mfg Cor
PO12025KG
2.5 kg
Poly Pail
Add to cart
18605310
View Documents
Spectrum Chemical Mfg Cor
PO12045KGBL
45 kg
Fiber Drum
Add to cart
Description
Description
Spectrum™ Chemical Polyethylene Glycol 1500, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
CAS
25322-68-3
Molecular Weight (g/mol)
62.07
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Molecular Formula
(C2H4O)n
MDL Number
MFCD01779601
IUPAC Name
ethane-1,2-diol
Specifications
pH
4.5 to 7.5
CAS Max %
1
MDL Number
MFCD01779601
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name
ethane-1,2-diol
Grade
NF
Packaging
Poly Pail
CAS
25322-68-3
Molecular Formula
(C2H4O)n
Quantity
2.5 kg
SMILES
[H]OCCO
Molecular Weight (g/mol)
62.07
Ignition Residue
0.001
Viscosity
26-33 cSt
Videos
SDS
missing translation for 'documents'