missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene Glycol 2,000, TCI America™
Supplier: TCI America P203425G
Specifications
Polyethylene Glycol 2,000 | |
55°C | |
(C2H4O)n | |
25 g | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
Crystalline Powder |
25322-68-3 | |
White | |
MFCD01779601 | |
PEG 2,000 | |
[H]OCCO | |
62.07 |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
PEG 2,000 | |
[H]OCCO |
Safety and Handling
EINECSNumber : (7)-0129
RTECSNumber : TQ4041000
TSCA : Yes