missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 400

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

$78.73 - $161.88

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 400
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2199230
View Documents
Thermo Scientific Chemicals
B2199230
250 g
Each for $78.73
Only null left
 
AAB219920B
View Documents
Thermo Scientific Chemicals
B219920B
1000 g
Each for $161.88
Only null left
 
Description

Description

Polyethylene glycol is used in a variety of pharmaceutical formulations. It finds application in Hewlett-Packard designjet printers as an ink solvent and a lubricant for the print heads. It is an important raw material for the manufacture of polyuretnae, PEG salts of lauric, oleic and stearic acids and latex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
(C2H4O)n
MFCD01779601
PEG 400
[H]OCCO

Specifications

5 to 7
4°C to 8°C
H(OCH2CH2)nOH
1.126
PEG 400
250 g
[H]OCCO
62.07
Odorless
Hygroscopic
25322-68-3
(C2H4O)n
MFCD01779601
14,7568
Miscible with water,acetone,alcohols,benzene,glycerin,glycols and aromatic hydrocarbons. Slightly miscible with aliphatic hydrocarbons.
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
240°C (464°F)
1.466
Polyethylene glycol 400
Videos
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c

H227-H303-H315-H320-H333-H335

RTECSNumber : TQ3675000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only