missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene Glycol 4500, NF, Spectrum™ Chemical
P1792, 25322-68-3, H(OCH2CH2)nOH
$259.85 - $5161.68
Chemical Identifiers
CAS | 25322-68-3 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 62.07 |
MDL Number | MFCD01779601 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
IUPAC Name | ethane-1,2-diol |
SMILES | [H]OCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18605337
|
Spectrum Chemical Mfg Cor
P1792500GM |
500 g | Poly Bottle |
Each for $259.85
|
|
||||
18605338
|
Spectrum Chemical Mfg Cor
P179225KG |
2.5 kg | Poly Pail |
Each for $786.24
|
|
||||
18605340
|
Spectrum Chemical Mfg Cor
P179212KG |
12 kg | Poly Pail |
Each for $2,698.38
|
|
||||
18605339
|
Spectrum Chemical Mfg Cor
P179245KGBL |
45 kg | Fiber Drum |
Each for $5,161.68
|
|
||||
Description
Spectrum™ Chemical Polyethylene Glycol 4500, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
[H]OCCO |
(C2H4O)n | |
MFCD01779601 | |
ethane-1,2-diol |
Specifications
4.5 to 7.5 | |
1 | |
MFCD01779601 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
NF | |
Poly Bottle |
25322-68-3 | |
(C2H4O)n | |
500 g | |
[H]OCCO | |
62.07 | |
0.001 | |
140-200 cSt |