Learn More
Polyethylene glycol 6,000, Thermo Scientific Chemicals
$51.14 - $584.38
Chemical Identifiers
CAS | 25322-68-3 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 62.07 |
MDL Number | MFCD01779601 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
IUPAC Name | ethane-1,2-diol |
SMILES | [H]OCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1754130
|
Thermo Scientific Chemicals
A1754130 |
250 g |
Each for $51.14
|
|
|||||
AAA175410B
|
Thermo Scientific Chemicals
A175410B |
1000 g |
Each for $146.10
|
|
|||||
AAA175410I
|
Thermo Scientific Chemicals
A175410I |
5000 g |
Each for $584.38
|
|
|||||
Description
Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In Gas chromatorgraphy, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
[H]OCCO |
(C2H4O)n | |
MFCD01779601 | |
ethane-1,2-diol |
Specifications
Powder | |
60°C to 63°C | |
MFCD01779601 | |
Soluble in water and aromatic hydrocarbons. Slightly and aliphatic hydrocarbons. | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
Polyethylene Glycol 6000 |
25322-68-3 | |
(C2H4O)n | |
14,7568 | |
250 g | |
[H]OCCO | |
62.07 |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
H315-H320
EINECSNumber : 203-473-3
RTECSNumber : TQ4100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only