Learn More
![](http://beta-static.fishersci.ca/images/Gray.gif)
Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
![Edge](/content/dam/fishersci/glyphs/edge.png)
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
Supplier: Fisher BioReagents FLBP2331
Description
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
PEG 8000 is suitable as a medium for the fusion of mammalian cells. It is also used to precipitate bacteriophage from lysed cell supernatants.Specifications
White | |
5 to 7 | |
25322-68-3 | |
(C2H4O)n | |
MFCD01779601 | |
PEG 8000, Polyethylene Oxide, Carbowax™ | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
7000 to 9000 | |
Poly Bottle |
Polyethylene Glycol 8000 | |
Solid | |
55°C | |
H(OCH2CH2)nOH | |
15, 7688 | |
1 kg | |
[H]OCCO | |
62.07 | |
Pass Test |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
PEG 8000, Polyethylene Oxide, Carbowax™ | |
[H]OCCO |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Gray.gif)
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0
Recommended Storage : RT