Learn More
Polyethylene glycol diacrylate, M.W. 8,000, Thermo Scientific Chemicals
$63.55 - $748.74
Chemical Identifiers
CAS | 26570-48-9 |
---|---|
Molecular Formula | C5H10O4 |
Molecular Weight (g/mol) | 134.131 |
MDL Number | MFCD00081876 |
InChI Key | FCDZZFLRLVQGEH-UHFFFAOYSA-N |
Synonym | acrylic acid; ethylene glycol, poly ethylene glycol diacrylate, polyethylene glycol diacrylate, poly ethylene glycol acrylate, average mn ∼375 |
PubChem CID | 16212859 |
IUPAC Name | ethane-1,2-diol;prop-2-enoic acid |
SMILES | C=CC(=O)O.C(CO)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA46801MD
|
Thermo Scientific Chemicals
046801MD |
250 mg |
Each for $63.55
|
|
|||||
AA4680103
|
Thermo Scientific Chemicals
04680103 |
1 g |
Each for $180.05
|
|
|||||
AA4680106
|
Thermo Scientific Chemicals
04680106 |
5 g |
Each for $748.74
|
|
|||||
Description
Polyethylene glycol diacrylate is used to prepare photopolymerized hydrogels which are used as scaffolds for tissue regeneration and other biological applications. It acts as an important raw material for the synthesis of poly(ethylene glycol) urethane-dimethacrylates (PEGUDM) by reacting with 2-isocyanatoethyl methacrylate. It contributes to chemical resistance, flexibility, adhesion, low shrinkage, abrasion resistance and impact strength.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26570-48-9 | |
134.131 | |
FCDZZFLRLVQGEH-UHFFFAOYSA-N | |
16212859 | |
C=CC(=O)O.C(CO)O |
C5H10O4 | |
MFCD00081876 | |
acrylic acid; ethylene glycol, poly ethylene glycol diacrylate, polyethylene glycol diacrylate, poly ethylene glycol acrylate, average mn ∼375 | |
ethane-1,2-diol;prop-2-enoic acid |
Specifications
White | |
26570-48-9 | |
MFCD00081876 | |
Miscible with water. | |
FCDZZFLRLVQGEH-UHFFFAOYSA-N | |
ethane-1,2-diol;prop-2-enoic acid | |
16212859 | |
Polyethylene glycol diacrylate, M.W. 8,000 |
Granular Solid | |
C5H10O4 | |
acrylic acid; ethylene glycol, poly ethylene glycol diacrylate, polyethylene glycol diacrylate, poly ethylene glycol acrylate, average mn ∼375 | |
250 mg | |
C=CC(=O)O.C(CO)O | |
134.131 | |
Light sensitive |
Safety and Handling
GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
H315-H317-H318-H335
EINECSNumber : 607-960-5
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only