Learn More
Polyethylene glycol methyl ether methacrylate, M.W. 5,000, Thermo Scientific Chemicals
$61.49 - $709.32
Chemical Identifiers
CAS | 26915-72-0 |
---|---|
Molecular Formula | C13H24O6 |
Molecular Weight (g/mol) | 276.329 |
MDL Number | MFCD00241432 |
InChI Key | KRCGBOKYIUDIFY-UHFFFAOYSA-N |
Synonym | 2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate |
PubChem CID | 93675 |
IUPAC Name | 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate |
SMILES | CC(=C)C(=O)OCCOCCOCCOCCOC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA46838MD
|
Thermo Scientific Chemicals
46838MD |
250 mg |
Each for $61.49
|
|
|||||
AA4683803
|
Thermo Scientific Chemicals
04683803 |
1 g |
Each for $173.42
|
|
|||||
AA4683806
|
Thermo Scientific Chemicals
04683806 |
5 g |
Each for $709.32
|
|
|||||
Description
Polyethylene glycol methyl ether methacrylate, is used as Emulsion polymers, cosmetics, Construction, emulsifiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26915-72-0 | |
276.329 | |
KRCGBOKYIUDIFY-UHFFFAOYSA-N | |
93675 | |
CC(=C)C(=O)OCCOCCOCCOCCOC |
C13H24O6 | |
MFCD00241432 | |
2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate | |
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate |
Specifications
White | |
26915-72-0 | |
MFCD00241432 | |
It is soluble in water. | |
KRCGBOKYIUDIFY-UHFFFAOYSA-N | |
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate | |
93675 |
Granular Solid | |
C13H24O6 | |
2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate | |
250 mg | |
CC(=C)C(=O)OCCOCCOCCOCCOC | |
276.329 | |
Polyethylene glycol methyl ether methacrylate, M.W. 5,000 |
Safety and Handling
GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
H315-H317-H318-H335
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only