missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol Monomethyl Ether 2000, TCI America™
Supplier: TCI America P2186100G
Specifications
| Polyethylene Glycol Monomethyl Ether 2000 | |
| 53°C | |
| (C2H4O)nCH4O | |
| 100 g | |
| XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
| 2-methoxyethan-1-ol | |
| 8019 | |
| Crystalline Powder |
| 9004-74-4 | |
| White | |
| MFCD00084416 | |
| methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
| COCCO | |
| 76.10 | |
| CHEBI:46790 |
Chemical Identifiers
| 9004-74-4 | |
| 76.10 | |
| XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
| 8019 | |
| 2-methoxyethan-1-ol |
| (C2H4O)nCH4O | |
| MFCD00084416 | |
| methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
| CHEBI:46790 | |
| COCCO |
Safety and Handling
EINECSNumber : (7)-0097
RTECSNumber : TR1581750
TSCA : Yes