missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene glycol monomethylether, 1,900, Thermo Scientific Chemicals
$82.23 - $342.49
Chemical Identifiers
CAS | 9004-74-4 |
---|---|
Molecular Formula | (C2H4O)nCH4O |
Molecular Weight (g/mol) | 76.10 |
MDL Number | MFCD00084416 |
InChI Key | XNWFRZJHXBZDAG-UHFFFAOYSA-N |
Synonym | methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em |
PubChem CID | 8019 |
ChEBI | CHEBI:46790 |
IUPAC Name | 2-methoxyethanol |
SMILES | COCCO |
Description
Polyethylene glycol monomethylether is used in the introduction of hydrophilic sites into polymers. It is also used to stabilize polymer emulsions. Further, it is employed in the synthesis of comb polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9004-74-4 | |
76.10 | |
XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
8019 | |
2-methoxyethanol |
(C2H4O)nCH4O | |
MFCD00084416 | |
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
CHEBI:46790 | |
COCCO |
Specifications
White to Yellow | |
9004-74-4 | |
(C2H4O)nCH4O | |
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
100 g | |
COCCO | |
76.10 | |
CHEBI:46790 | |
Polyethylene glycol monomethylether 1,900 |
Solid Mass | |
52°C | |
MFCD00084416 | |
Slightly miscible with water. | |
XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
2-methoxyethanol | |
8019 | |
>110°C (230°F) |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only