Learn More
Polyethyleneimine, branched, M.W. 2,000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04052814
Description
Polyethyleneimine is used for various applications in products like detergents, adhesives, water treatment agents and cosmetics. It is also used as flocculating agent, enzyme carrier, wet-strength agent in the paper-making process, chelating agent form complexes with metal ions such as zinc and zirconium. It acts as scavenger for aldehydes and oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless to Yellow | |
M.W. 1,800 | |
9002-98-6 | |
MFCD00084427 | |
Soluble in hot water at a low pH,in methanol,ethanol,or chloroform,but insoluble in cold water,benzene,ethyl ether,and acetone. | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
aziridine | |
9033 | |
99% |
Polyethyleneimine, branched | |
Liquid | |
((C2H5N)zC2H4N)y(C2H5N)x | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
25 g | |
*-CCNCCN(-*)CCN-* | |
43.07 | |
CHEBI:30969 | |
Amine-like |
Chemical Identifiers
9002-98-6 | |
43.07 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
9033 | |
aziridine |
((C2H5N)zC2H4N)y(C2H5N)x | |
MFCD00084427 | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
CHEBI:30969 | |
*-CCNCCN(-*)CCN-* |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only