missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polymethylhydrosiloxane, Thermo Scientific Chemicals
$107.44 - $249.73
Chemical Identifiers
CAS | 9004-73-3 |
---|---|
Molecular Formula | (CH4OSi)n |
Molecular Weight (g/mol) | 60.13 |
MDL Number | MFCD00084478 |
InChI Key | TXYXVOOPRILIHR-UHFFFAOYSA-N |
Synonym | polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3 |
PubChem CID | 12487658 |
IUPAC Name | 2-methyl-1,2$l^{3}-oxasilinane |
SMILES | C[SiH](-*)O-* |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC175091000
|
Thermo Scientific Chemicals
175091000 |
100 g | Glass bottle |
Each for $107.44
|
|
||||
AC175095000
|
Thermo Scientific Chemicals
175095000 |
500 g | Glass bottle |
Each for $249.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
9004-73-3 | |
60.13 | |
TXYXVOOPRILIHR-UHFFFAOYSA-N | |
12487658 | |
C[SiH](-*)O-* |
(CH4OSi)n | |
MFCD00084478 | |
polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3 | |
2-methyl-1,2$l^{3}-oxasilinane |
Specifications
Colorless | |
9004-73-3 | |
(CH3)3SiO[(CH3)HSiO]nSi(CH3)3 | |
1.0000g/mL | |
Solubility in water: practically insoluble. Other solubilities: may react, soluble in most common organic solvents, (should not be used in halocarbon solvent because, of the possibility of hydride-halide interchange, on catalysis or with heating) | |
TXYXVOOPRILIHR-UHFFFAOYSA-N | |
2-methyl-1,2$l^{3}-oxasilinane | |
12487658 | |
Authentic | |
1.3970 to 1.4010 | |
15-30 mm2/s (25°C) |
Viscous Liquid | |
(CH4OSi)n | |
MFCD00084478 | |
polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3 | |
100 g | |
C[SiH](-*)O-* | |
60.13 | |
121°C | |
Glass bottle | |
1 | |
Polymethylhydrosiloxane |
RUO – Research Use Only