missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals

$138.05 - $293.25

Chemical Identifiers

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC196950010
SDS View Documents
Thermo Scientific Chemicals
196950010
1 kg Plastic bottle N/A
 
AC196950025
SDS View Documents
Thermo Scientific Chemicals
196950025
2.5 kg Plastic bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8017-16-1
MFCD00084480
Polyphosphoric acid
(HO3P)n
NBIIXXVUZAFLBC-UHFFFAOYSA-N
[H]OP(O)(O)=O

Specifications

8017-16-1
2.0500g/mL
2.05
(HO3P)n
MFCD00084480
NBIIXXVUZAFLBC-UHFFFAOYSA-N
Polyphosphoric acid
Pure
550.0°C
Plastic bottle
1 kg
Viscous Liquid
Solubility in water: may decompose. Other solubilities: insoluble in hydrocarbons and halogenated hydrocar, soluble in alcohols
[H]OP(O)(O)=O
≥82% (as P2O5) (Alkalimetry)
Polyphosphoric acid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : May darken over time

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only