missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Polypropylene Burette Clamp
Eisco™ Polypropylene Burette Clamp feature a spring-loaded jack and two tapered wedges to assure a snug and safe fit.
Supplier: Eisco™ CH0262A
Description
- Single sided burette clamp
- Made with durable, corrosion resistant polypropylene
- Accommodates rod sizes 10 mm and 12.5 mm with 2 tapered wedges
- Spring-loaded jack tightly secures burette up to 15mm in diameter
Warranty
1 year limited manufacturer warranty
Specifications
14 mm | |
Polypropylene | |
Buret Clamp |
Burettes | |
Holding Burets with rod sizes 10 mm and 12.5 mm |