missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Polypropylene Conical Containers

Quality sampling tools for secure sample retrieval and shipment

Supplier:  Corning™ PC100004

 View more versions of this product

Catalog No. 07-201-939

Explore available promotions

Description

Description

  • Lightweight and break-resistant
  • Transparent or white Polypropylene (PP)
  • Assembled or non-assembled
Specifications

Specifications

Yes
Sterile
1 L
Snap Cap
105 mm
80/Cs.
Polypropylene
33.8 oz.
White
White
130 mm
80/Pk.
Videos
SDS
Documents

Documents

Promotions

Promotions