missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Polypropylene Conical Containers

Quality sampling tools for secure sample retrieval and shipment

Supplier:  Corning™ PC40009

 View more versions of this product

Catalog No. 07-201-987

Explore available promotions

Description

Description

  • Lightweight and break-resistant
  • Transparent or white Polypropylene (PP)
  • Assembled or non-assembled
Specifications

Specifications

Yes
Non-sterile
400 mL
LDPE
85 mm
185/Cs.
Polypropylene
6.7 oz.
Natural
Natural
100 mm
185/Pk.
Videos
SDS
Documents

Documents

Promotions

Promotions