Corning™ Polypropylene Conical Containers
Quality sampling tools for secure sample retrieval and shipment
Supplier:  Corning™ PC40005
 View more versions of this product
Catalog No. 07-901-945
Explore available promotions
Description
Description
  • Lightweight and break-resistant
  • Transparent or white Polypropylene (PP)
  • Assembled or non-assembled
Specifications
Specifications
Polypropylene
13.5 oz.
White
85 mm
185/Pk.
Sterile
400 mL
White
185/Cs.