missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Polypropylene Conical Flasks with Screw Caps

Eisco™ Polypropylene Conical Flasks with Screw Caps feature a rigid, durable, and translucent body.

$10.53 - $17.03

Specifications

Closure Color Clear
Closure Material Polypropylene
Color Clear
Material Polypropylene
For Use With (Application) Heating and holding liquids
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S89384
View Documents
Eisco™
CH0431A
100 mL
Each for $10.53
 
S13966
View Documents
Eisco™
CH0431B
250 mL
Each for $17.03
 
Description

Description

  • Plastic conical flask with screw cap
  • Durable polypropylene construction
  • Autoclavable
Specifications

Specifications

Clear
Clear
Heating and holding liquids
Autoclavable
No
Non-sterile
Screw Cap
Polypropylene
Polypropylene
Conical Flask
Reusable
Narrow
Flat
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'