missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polypropylene Disposable Beakers
Disposable or steam-autoclavable for reuse.
Supplier: Customer Channels Group (Ccg) L1240EMD
Description
- Resistant to most acids, solvents and other common reagents
- Special “dripless” lip
- Rib construction permits stacking without binding
- Graduated markings in mL and oz.
Specifications
150 mL | |
Spout | |
Beaker | |
Single-use |
Polypropylene | |
5 mL | |
Autoclavable | |
Yes |