Polypropylene Disposable Beakers
Disposable or steam-autoclavable for reuse.
Supplier:  Customer Channels Group (Ccg) L1260EMD
 View more versions of this product
Catalog No. S63362
Explore available promotions
Description
Description
  • Resistant to most acids, solvents and other common reagents
  • Special “dripless” lip
  • Rib construction permits stacking without binding
  • Graduated markings in mL and oz.
Specifications
Specifications
400 mL
Spout
Beaker
Single-use
Polypropylene
25 mL
Autoclavable
Yes