missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene glycol 2000

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

$82.08 - $141.95

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL14699AE
View Documents
Thermo Scientific Chemicals
L14699AE
100 mL
Each for $82.08
Only null left
 
AAL14699AP
View Documents
Thermo Scientific Chemicals
L14699AP
500 mL
Each for $141.95
Only null left
 
Description

Description

Used to make polyester compounds, absorb extra water and maintain moisture in certain medicines, cosmetics, or food products and as a base for deicing solutions. Propylene glycol is used by the chemical, food, and pharmaceutical industries as an antifreeze. It is a solvent for food colors and flavors, and in the paint and plastics industries. Propylene glycol is also used to create artificial smoke or fog used in fire-fighting training and in theatrical productions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25322-69-4
134.18
DNIAPMSPPWPWGF-UHFFFAOYNA-N
32881
[H]OC(C)CO
(C3H6O)n
MFCD00084446
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
2-(2-hydroxypropoxy)propan-1-ol

Specifications

25322-69-4
MFCD00084446
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
100 mL
[H]OC(C)CO
134.18
2000
1.451
(C3H6O)n
1.002
Miscible with water,ethanol,diethyl ether and chloroform.
DNIAPMSPPWPWGF-UHFFFAOYNA-N
2-(2-hydroxypropoxy)propan-1-ol
32881
194°C (381°F)
Polypropylene glycol 2000
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 500-039-8

RTECSNumber : TR6130000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only