missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polypropylene glycol 3,000, Thermo Scientific Chemicals

$54.96 - $213.66

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4081418
View Documents
Thermo Scientific Chemicals
04081418
50 g
Each for $54.96
 
AA4081430
View Documents
Thermo Scientific Chemicals
04081430
250 g
Each for $213.66
 
Description

Description

Propylene glycol is used in the plastics industry for the manufacture of polyester fibers and alkyd resins. It is used as a main ingredient in automobile antifreeze and engine-cooling liquids and in brake and hydraulic fluids. It is used as a humectant in foods and cosmetics. It is used as a solvent for coloring or flavoring agents as well as in many oral, injectable or topical pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25322-69-4
134.18
DNIAPMSPPWPWGF-UHFFFAOYNA-N
32881
[H]OC(C)CO
(C3H6O)n
MFCD00084446
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
2-(2-hydroxypropoxy)propan-1-ol

Specifications

Liquid
(C3H6O)n
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
50 g
[H]OC(C)CO
134.18
Polypropylene glycol 3,000
25322-69-4
MFCD00084446
Soluble in ethyl alcohol,ethyl ether.
DNIAPMSPPWPWGF-UHFFFAOYNA-N
2-(2-hydroxypropoxy)propan-1-ol
32881
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only