Globe Scientific Polypropylene Test Tubes with Screw Cap
Multipurpose screw cap test tubes are offered in polystyrene (PS) or polypropylene (PP) and suit variety of applications.
Supplier:  Globe Scientific 6180
Catalog No. 22-010-063
Explore available promotions
Description
Description
  • Designed to be leak-tight allowing tubes to be used for storage or transport
  • Screw caps are polyethylene
  • Polypropylene tubes are autoclavable
  • Round bottom
Specifications
Specifications
Natural
Natural
100 mm
Attached screw cap
Autoclavable
Test Tube
Polyethylene
12 mL
Polypropylene
Non-sterile
No