InterLab™ Supply Polyseed™ BOD and Polyseed-NX™ CBOD Capsules
Produce fast, economical BOD analyses with consistent results. Available in two formulations.
Supplier:  InterLab™ Supply P210
 View more versions of this product
Catalog No. 13-297-201
Explore available promotions
Description
Description
  • EPA accepted
  • Each capsule contains specialized microbial cultures for use in a broad range of areas involving degradation of industrial and municipal waste
  • To use, mix capsule contents with 0.5L of APHA standard dilution water at 20°C (68°F), stir for at least 60 minutes
  • Mixture yields seed inoculum for up to 200 BOD tests
  • For best results, use within one year of purchase
  • Also available as Polyseed-NX, which contains an approved chemical additive for the inhibition of ammonia nitrogen (NH3-N) in wastewater samples for CBOD analysis
Order Info
Supplied as a bottle of 50 capsules.
Specifications
Specifications
EPA
Specialized microbial cultures
Polyseed-NX CBOD Capsules
Standard seed source for the degradation of both industrial and municipal waste in CBOD testing
Up to 200