missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 40
CAS: 9005-66-7 | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | 522.68 g/mol
$43.70 - $143.07
Chemical Identifiers
| CAS | 9005-66-7 |
|---|---|
| Molecular Formula | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 |
| Molecular Weight (g/mol) | 522.68 |
| MDL Number | MFCD00165345 |
| InChI Key | DVRFSZFBSOKHQW-UHFFFAOYNA-N |
| Synonym | Polyoxyethylene (20) Sorbitan Monopalmitate, Polysorbate 40 |
| PubChem CID | 92329579 |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC334140250
|
Thermo Scientific Chemicals
334140250 |
25 mL | Glass bottle |
Each for $43.70
|
|
||||
|
AC334142500
|
Thermo Scientific Chemicals
334142500 |
250 mL | Glass bottle |
Each for $97.34
|
|
||||
|
AC334140010
|
Thermo Scientific Chemicals
334140010 |
1 L | Glass bottle |
Each for $143.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9005-66-7 | |
| 522.68 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| 92329579 |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| MFCD00165345 | |
| Polyoxyethylene (20) Sorbitan Monopalmitate, Polysorbate 40 | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Specifications
| Yellow | |
| Viscous Liquid or Paste At 25°C | |
| 25 mL | |
| 89 to 105mg KOH/g | |
| Glass bottle | |
| 1.05 | |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| Polyoxyethylene (20) Sorbitan Monopalmitate, Polysorbate 40 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
| 92329579 |
| 9005-66-7 | |
| 1.0500g/mL | |
| >200°C | |
| Authentic | |
| 41 to 52mg KOH/g | |
| ca.200 mPa.s (50°C) | |
| MFCD00165345 | |
| Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, xylene, insoluble in propylene glycol and mineral oil | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 | |
| Polysorbate 40 |
RUO – Research Use Only