missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 80
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol
$81.25 - $1276.85
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC278632500
|
Thermo Scientific Chemicals
278632500 |
250 mL |
Each for $81.25
|
|
|||||
|
AC278630010
|
Thermo Scientific Chemicals
278630010 |
1 L |
Each for $177.54
|
|
|||||
|
AC278630025
|
Thermo Scientific Chemicals
278630025 |
2.5 L |
Each for $390.44
|
|
|||||
|
AC278630100
|
Thermo Scientific Chemicals
278630100 |
10 L |
Each for $1,276.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9005-65-6 | |
| Polyoxyethylene(20)sorbitan monooleate |
Specifications
| 9005-65-6 | |
| Indoors +20°C | |
| 250 mL,250 mL | |
| MFCD00082107 | |
| HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
| 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate | |
| Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, 10g/L xylene, insoluble in propylene glycol and mineral oil | |
| Viscous Liquid |
| Yellow to Amber | |
| 1.0800 g/mL | |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
| Glass bottle | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 604.82 | |
| Polyoxyethylene(20)sorbitan monooleate | |
| Polysorbate 80 |
RUO – Research Use Only