Cole-Parmer™ Essentials Pipette Basins
V-shaped design minimizes reagent waste
Supplier:  Cole-Parmer™ Essentials 0439529
 View more versions of this product
Catalog No. 03-391-541
Explore available promotions
Description
Description
  • Certified RNase-, DNase-, and pyrogen-free
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. They are available in a variety of volumes come in either sterile or nonsterile formats.
Single Chamber Reservoirs are made of white polystyrene and constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring. Available in 25, 50, and 100 mL volumes, the reservoirs are ideal for 8- and 12-channel pipettors.
The 12-Chamber Reservoir is designed for serial dilutions or pipetting different reagents using a 12-channel pipettor. Each numbered chamber holds up to 3 mL. The chemical-resistant polypropylene reservoir can withstand temperatures from -122 to 250°F (-80 to 121°C). Tight fitting lid minimizes evaporation or contamination during incubation or storage.
Specifications
Specifications
White
Polystyrene
Pipetting Reservoir
Individually Wrapped
100 mL
Sterile
50