missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Polystyrene Sign: CAUTION - HOT SURFACE. DO NOT TOUCH.
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$13.59 - $20.08
Specifications
Material | Polystyrene |
---|---|
Shape | Rectangular |
Displayed Text | CAUTION - HOT SURFACE. DO NOT TOUCH. |
Color | Black on White |
Catalog Number | Mfr. No. | Dimensions (L x W) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Includes | Price | Quantity | ||||
19-110-828
|
Brady™
21788 |
10 x 7 in. | Hand/Flame Picto |
Each for $13.59
|
|
||||
19-110-715
|
Brady™
143822 |
14 x 10 in. | Heat Picto |
Each for $20.08
|
|
||||
19-110-712
|
Brady™
143819 |
10 x 7 in. | Heat Picto |
Each for $13.59
|
|
||||
Description
- Polystyrene material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Polystyrene signs are thin, pliable and durable
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Polystyrene | |
Rectangular | |
CAUTION - HOT SURFACE. DO NOT TOUCH. | |
Black on White |