Brady™ Polystyrene Sign: (NO TEXT), White
Improve workplace safety with proper signage and markings. This blank Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Supplier:  Brady™ 25376
 View more versions of this product
Catalog No. 19-104-734
Explore available promotions
Description
Description
  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Polystyrene
10 x 7 in.
White
Rectangular
Blank
Product Suggestions
Product Suggestions