missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heathrow Scientific™ Polystyrene Sterile Single Wrapped Reagent Reservoir
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
$128.74 - $513.56
Specifications
Color | White |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | Single Wrapped |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | ||||||
03449255
|
Heathrow Scientific™
120637 |
5 mL |
Pack of 50 Each for $128.74
|
|
||||||
03449257
|
Heathrow Scientific™
120639 |
10 mL |
Pack of 100 Each for $238.26
|
|
||||||
03449259
|
Heathrow Scientific™
120641 |
25 mL |
Pack of 100 Each for $403.09
|
|
||||||
03449262
|
Heathrow Scientific™
120644 |
50 mL |
Pack of 100 Each for $370.62
|
|
||||||
03449278
|
Heathrow Scientific™
120750 |
55 mL |
Pack of 100 Each for $513.56
|
|
||||||
03449266
|
Heathrow Scientific™
120648 |
100 mL |
Pack of 100 Each for $513.56
|
|
||||||
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sterile, sealed in bags and electron beam sterilized
- DNase, RNase and Pyrogen free
- Sealed in easy-to-open bags
- Quality certificate available on request
- Disposable
Specifications
White | |
Sterile | |
Single Wrapped | |
1 | |
Convoluted |
Polystyrene | |
Reagent Reservoir | |
Not Autoclavable | |
Single-use |