missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyvinyl alcohol, 86-89% hydrolyzed, medium molecular weight, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04123936

 View more versions of this product

Catalog No. AA4123936


Explore available promotions

Description

Description

Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Polyvinyl alcohol
Powder
(C2H4O)n
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
500 g
OC(-*)C-*
44.05
86 to 89%
Medium molecular weight, Hydrolyzed
9002-89-5
MFCD00081922
Soluble in water.
IMROMDMJAWUWLK-UHFFFAOYSA-N
ethenol
11199
Odorless

Chemical Identifiers

9002-89-5
44.05
IMROMDMJAWUWLK-UHFFFAOYSA-N
11199
OC(-*)C-*
(C2H4O)n
MFCD00081922
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
ethenol
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only