Learn More
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04124336
Description
Polyvinyl alcohol is used in food industries as a binding and coating agent. Its viscosity favors the application as coating agent to tablets and capsules. Mixed-matrix membrane prepared by dispersion of NaA zeolite particles is used to study the pervaporative separation of water-butanol mixtures. Polyvinyl alcohol /gelatin based biocompatible polymeric scaffolds have been developed for 3D cancer models.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polyvinyl alcohol | |
Solid | |
(C2H4O)n | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
500 g | |
OC(-*)C-* | |
44.05 | |
98 to 99% |
High molecular weight, Hydrolyzed | |
9002-89-5 | |
MFCD00081922 | |
Soluble in water. | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
ethenol | |
11199 | |
Odorless |
Chemical Identifiers
9002-89-5 | |
44.05 | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
11199 | |
OC(-*)C-* |
(C2H4O)n | |
MFCD00081922 | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
ethenol |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only