missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyvinylpyrrolidone, average M.W. 3500, K12, Thermo Scientific Chemicals

$342.98 - $342.98

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC276142500
SDS View Documents
Thermo Scientific Chemicals
276142500
250 g Plastic bottle
Each for $342.98
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9003-39-8
111.14
WHNWPMSKXPGLAX-UHFFFAOYSA-N
6917
1-ethenylpyrrolidin-2-one
(C6H9NO)n
MFCD01076626
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
CHEBI:82551
*-CC(-*)N1CCCC1=O

Specifications

White to White-Yellow
Powder or Flakes
(C6H9NO)n
15, 7814
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether
WHNWPMSKXPGLAX-UHFFFAOYSA-N
1-ethenylpyrrolidin-2-one
6917
11.5 to 12.8% (nitrogen, on anh. subst.), 10ppm 1-vinylpyrrolidin-2-one max. (HPLC), 3.0% max. 2-pyrrolidone (HPLC)
Authentic
5% Max. (K.F.)
3.0 to 5.0 (5% aq. soln.)
9003-39-8
MFCD01076626
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
250 g
*-CC(-*)N1CCCC1=O
111.14
CHEBI:82551
Passes Test
Plastic bottle
Polyvinylpyrrolidone, K12
Videos
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

HYGROSCOPIC

RTECSNumber : TR8250000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only