missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone-iodine complex

CAS: 25655-41-8 | (C6H9NO)nI2 | 364.95 g/mol

$203.27 - $3143.68

Chemical Identifiers

CAS 25655-41-8
Molecular Formula (C6H9NO)nI2
Molecular Weight (g/mol) 364.95
MDL Number MFCD00084483
InChI Key CPKVUHPKYQGHMW-UHFFFAOYSA-N
Synonym povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
PubChem CID 410087
IUPAC Name 1-ethenylpyrrolidin-2-one;molecular iodine
SMILES II.*-CC(-*)N1CCCC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC229471000
SDS View Documents
Thermo Scientific Chemicals
229471000
100 g
Each for $203.27
Only null left
 
AC229470025
SDS View Documents
Thermo Scientific Chemicals
229470025
2.5 kg
Each for $3,143.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25655-41-8
364.95
CPKVUHPKYQGHMW-UHFFFAOYSA-N
410087
II.*-CC(-*)N1CCCC1=O
(C6H9NO)nI2
MFCD00084483
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
1-ethenylpyrrolidin-2-one;molecular iodine

Specifications

25655-41-8
Brown-Red to Yellow
Authentic
(C6H9NO)nI2
MFCD00084483
15, 7815
Solubility in water: soluble. Other solubilities: soluble in ethanol,isopropanol,glycerol;,practically insoluble in acetone
II.*-CC(-*)N1CCCC1=O
364.95
9.0 to 12.0% (available iodine) (on dried substance)
Polyvinylpyrrolidone-iodine complex
300°C
8% max. (5 g, 105°C)
Plastic bottle
(C6H9NO)n·xI
100 g
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
CPKVUHPKYQGHMW-UHFFFAOYSA-N
1-ethenylpyrrolidin-2-one;molecular iodine
410087
Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TR1579600

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only