missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ponceau S, MP Biomedicals

Supplier:  MP Biomedicals Inc 0215194280

 View more versions of this product

Catalog No. ICN15194280


Explore available promotions

Description

Description

Specifications

Specifications

Ponceau S
∽ 75%
3-Hydroxy-4-[2-sulfo-4-(4-sulfo- phenylazo)phenylazo]-2, 7- naphthalene-disulfonic acid tetrasodium salt
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
760.552
760.6
Powder
6226-79-5
C22H12N4Na4O13S4
VSXKEUCERCWGKF-STNZDNLRSA-J
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
11320219
100 g

Chemical Identifiers

6226-79-5
760.552
3-Hydroxy-4-[2-sulfo-4-(4-sulfo- phenylazo)phenylazo]-2, 7- naphthalene-disulfonic acid tetrasodium salt
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
C22H12N4Na4O13S4
VSXKEUCERCWGKF-STNZDNLRSA-J
11320219
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature.

SDS
Documents

Documents

Promotions

Promotions