missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Ponceau S Staining Solution
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.
$433.20 - $433.20
Specifications
Detection Method | Colorimetric |
---|---|
Formulation | 0.1% (w/v) in 5% acetic acid |
For Use With (Application) | Western Blotting |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
A40000278X3
|
Thermo Scientific™
A40000278X3 |
Ponceau S Staining Solution, 3 x 1000 mL |
Each for $433.20
|
|
|||||
Description
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein
Specifications
Colorimetric | |
0.1% (w/v) in 5% acetic acid | |
Western Blotting |