CoorsTek™ Porcelain Coal Moisture Testing Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
$72.03 - $72.03
Specifications
Material Porcelain
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
08-103D
Accessories View Documents
CoorsTek™
CRS66054DUP
35 mL
Each for $72.03
Case of 24 Each for $1,728.72
Description
Description
  • Porcelain with rounded bottom
  • Glazed inside and out except on rim
  • 20mL size recommended for ASTM™ D 3176
  • Capsule form; without covers
Specifications
Specifications
Porcelain
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'