ICU Medical Portex™ Line Draw Plus Arterial Blood Sampling Kits DFS Item
Line Draw Plus aspirating syringes allow for blood sampling directly from an arterial line. The syringes use an advanced formula of dry lithium heparin for consistent and accurate readings. They are available with the following configurations: 1 mL Luer Slip Syringe, 3 mL Luer Lock Syringe, or 3 mL Luer Slip Slip Syringe.
Supplier:  ICU Medical 4042LH
 View more versions of this product
Catalog No. 22-024-958
Explore available promotions
Description
Description
Advanced formula offsets both binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level. Provides consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactate.
  • Umbilical and arterial syringes contain a 23.5 IU/mL concentration of dry lithium heparin
  • Low-heparin arterial syringes have a 7 IU/mL concentration of dry lithium heparin
  • Anaerobic syringes contain 23.5 IU/mL of electrolyte-balanced liquid sodium heparin
†With support kit (alcohol prep pad, gauze pad, biohazard-labeled ice bag).
Specifications
Specifications
Allow for blood sampling directly from an arterial line, provides consistent and accurate analysis for all parameters tested
Arterial Blood Sampling Kit
Polybag
3mL line draw luer-lock syringe with Filter-Pro™ device, 7 IU/mL concentration of low dry lithium heparin as anticoagulant
200/Cs.
3 mL