missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium 2,4,5-Trichlorophenoxyacetate 98.0+%, TCI America™
Supplier: TCI America T1509500G
Specifications
Potassium 2,4,5-Trichlorophenoxyacetate | |
White | |
MFCD00191590 | |
2,4,5-Trichlorophenoxyacetic Acid Potassium Salt, 2,4,5-T Potassium Salt | |
C1=C(C(=CC(=C1Cl)Cl)Cl)OCC(=O)[O-].[K+] | |
293.565 | |
293.57 | |
Crystalline Powder |
37785-57-2 | |
C8H4Cl3KO3 | |
500 g | |
LUDUIQAQFKIYND-UHFFFAOYSA-M | |
potassium;2-(2,4,5-trichlorophenoxy)acetate | |
23684557 | |
≥98.0% (T) |
Chemical Identifiers
37785-57-2 | |
293.565 | |
LUDUIQAQFKIYND-UHFFFAOYSA-M | |
23684557 | |
C1=C(C(=CC(=C1Cl)Cl)Cl)OCC(=O)[O-].[K+] |
C8H4Cl3KO3 | |
MFCD00191590 | |
2,4,5-Trichlorophenoxyacetic Acid Potassium Salt, 2,4,5-T Potassium Salt | |
potassium;2-(2,4,5-trichlorophenoxy)acetate |
Safety and Handling
TSCA : No