Learn More
Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03916304
Description
Used to convert the tert-butylammmonium salt of clavulanic acid into potassium clavulanate or clavulanate potassium. Also used as a catalyst for polyurethane systems (foams) and for unsaturated polyester resin systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium 2-ethylhexanoate | |
Characteristic | |
C8H15KO2 | |
MFCD00045896 | |
UN1993 | |
Partly miscible with water. | |
[K+].CCCCC(CC)C([O-])=O | |
182.30 | |
182.3 | |
75% w/w soln. |
3164-85-0 | |
(metals basis) | |
K[OOCCH(C2H5)C4H9] | |
2 g | |
potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p | |
ZUFQCVZBBNZMKD-UHFFFAOYNA-M | |
potassium;2-ethylhexanoate | |
23669619 | |
99.9% | |
Viscous Liquid |
Chemical Identifiers
3164-85-0 | |
182.30 | |
ZUFQCVZBBNZMKD-UHFFFAOYNA-M | |
[K+].CCCCC(CC)C([O-])=O |
C8H15KO2 | |
MFCD00045896 | |
23669619 |
Safety and Handling
GHS H Statement
H226-H361-H373
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
H315-H318-H361
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 221-625-7
TSCA : Yes
Recommended Storage : Ambient temperatures
For Manufacturing and Laboratory Use. Not for beverage use