missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium (2-Methoxyphenyl)trifluoroborate 98.0+%, TCI America™

Supplier:  TCI America P18101G

 View more versions of this product

Catalog No. P18101G


Explore available promotions

Description

Description

Specifications

Specifications

Potassium (2-Methoxyphenyl)trifluoroborate
White
MFCD04115756
potassium 2-methoxyphenyl trifluoroborate, potassium trifluoro 2-methoxyphenyl borate, potassium 2-methoxyphenyltrifluoroborate, potassium trifluoro 2-methoxyphenyl boranuide, pubchem11466, amtb101, trifluoro 2-methoxyphenyl potassioboron v, potassium trifluoro-2-methoxyphenyl boranuide, potassium trifluoro 2-methoxyphenyl borate 1-
[K+].COC1=CC=CC=C1[B-](F)(F)F
214.04
214.04
Crystalline Powder
236388-46-8
C7H7BF3KO
1 g
IXJPOOXJHKETAZ-UHFFFAOYSA-N
potassium trifluoro(2-methoxyphenyl)boranuide
23684966
≥98.0% (HPLC)

Chemical Identifiers

236388-46-8
214.04
IXJPOOXJHKETAZ-UHFFFAOYSA-N
23684966
[K+].COC1=CC=CC=C1[B-](F)(F)F
C7H7BF3KO
MFCD04115756
potassium 2-methoxyphenyl trifluoroborate, potassium trifluoro 2-methoxyphenyl borate, potassium 2-methoxyphenyltrifluoroborate, potassium trifluoro 2-methoxyphenyl boranuide, pubchem11466, amtb101, trifluoro 2-methoxyphenyl potassioboron v, potassium trifluoro-2-methoxyphenyl boranuide, potassium trifluoro 2-methoxyphenyl borate 1-
potassium trifluoro(2-methoxyphenyl)boranuide
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions