missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-methoxypyridine-5-trifluoroborate, 97%, Thermo Scientific™

$85.97 - $376.01

Chemical Identifiers

CAS 1144573-61-4
Molecular Formula C6H6BF3KNO
Molecular Weight (g/mol) 215.024
MDL Number MFCD09993111
InChI Key ASWXYOAUUZSFCK-UHFFFAOYSA-N
Synonym potassium 2-methoxypyridine-5-trifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl borate, amtb168, potassium 6-methoxy-3-pyridyltrifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl boranuide
PubChem CID 72698470
IUPAC Name potassium;trifluoro-(6-methoxypyridin-3-yl)boranuide
SMILES [B-](C1=CN=C(C=C1)OC)(F)(F)F.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6164503
View Documents
Thermo Scientific Chemicals
H6164503
1 g
Each for $85.97
Only null left
 
AAH6164506
View Documents
Thermo Scientific Chemicals
H6164506
5 g
Each for $376.01
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1144573-61-4
215.024
ASWXYOAUUZSFCK-UHFFFAOYSA-N
72698470
[B-](C1=CN=C(C=C1)OC)(F)(F)F.[K+]
C6H6BF3KNO
MFCD09993111
potassium 2-methoxypyridine-5-trifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl borate, amtb168, potassium 6-methoxy-3-pyridyltrifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl boranuide
potassium;trifluoro-(6-methoxypyridin-3-yl)boranuide

Specifications

1144573-61-4
MFCD09993111
potassium 2-methoxypyridine-5-trifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl borate, amtb168, potassium 6-methoxy-3-pyridyltrifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl boranuide
[B-](C1=CN=C(C=C1)OC)(F)(F)F.[K+]
215.024
215.02
Potassium 2-methoxypyridine-5-trifluoroborate
C6H6BF3KNO
1 g
ASWXYOAUUZSFCK-UHFFFAOYSA-N
potassium;trifluoro-(6-methoxypyridin-3-yl)boranuide
72698470
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'