missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium 2-methylthiophenyltrifluoroborate, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5258703

 View more versions of this product

Catalog No. AAH5258703


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Potassium 2-methylthiophenyltrifluoroborate
274°C to 276°C
MFCD04115773
potassium trifluoro 2-methylthio phenyl borate, potassium 2-methylthiophenyl trifluoroborate, potassium trifluoro 2-methylsulfanyl phenyl boranuide, potassium 2-methylsulfanylphenyl trifluoroborate, potassium 2-methylthiophenyltrifluoroborate, potassium trifluoro-2-methylthio phenyl boranuide, potassium trifluoro 2-methylsulfanyl phenyl borate 1-, potassium ion trifluoro 2-methylsulfanyl phenyl boranuide, potassium tris fluoranyl-2-methylsulfanylphenyl boranuide
[B-](C1=CC=CC=C1SC)(F)(F)F.[K+]
230.097
230.1
850623-77-7
C7H7BF3KS
1 g
ACOYJOWMHXJLJP-UHFFFAOYSA-N
potassium;trifluoro-(2-methylsulfanylphenyl)boranuide
44717213
96%

Chemical Identifiers

850623-77-7
230.097
ACOYJOWMHXJLJP-UHFFFAOYSA-N
44717213
[B-](C1=CC=CC=C1SC)(F)(F)F.[K+]
C7H7BF3KS
MFCD04115773
potassium trifluoro 2-methylthio phenyl borate, potassium 2-methylthiophenyl trifluoroborate, potassium trifluoro 2-methylsulfanyl phenyl boranuide, potassium 2-methylsulfanylphenyl trifluoroborate, potassium 2-methylthiophenyltrifluoroborate, potassium trifluoro-2-methylthio phenyl boranuide, potassium trifluoro 2-methylsulfanyl phenyl borate 1-, potassium ion trifluoro 2-methylsulfanyl phenyl boranuide, potassium tris fluoranyl-2-methylsulfanylphenyl boranuide
potassium;trifluoro-(2-methylsulfanylphenyl)boranuide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions