missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium 3,5-Diiodosalicylate 97.0+%, TCI America™
Supplier: TCI America D16861G
Specifications
Potassium 3,5-Diiodosalicylate | |
White-Yellow | |
MFCD00058984 | |
3,5-Diiodosalicylic Acid Potassium Salt | |
C1=C(C=C(C(=C1I)O)C(=O)[O-])I.[K+] | |
428.005 | |
428.01 | |
Crystalline Powder |
1015937-56-0 | |
C7H3I2KO3 | |
1 g | |
SNHIVWOGVZUVLO-UHFFFAOYSA-M | |
potassium;2-hydroxy-3,5-diiodobenzoate | |
23716296 | |
≥97.0% (T) |
Chemical Identifiers
1015937-56-0 | |
428.005 | |
SNHIVWOGVZUVLO-UHFFFAOYSA-M | |
23716296 | |
C1=C(C=C(C(=C1I)O)C(=O)[O-])I.[K+] |
C7H3I2KO3 | |
MFCD00058984 | |
3,5-Diiodosalicylic Acid Potassium Salt | |
potassium;2-hydroxy-3,5-diiodobenzoate |
Safety and Handling
TSCA : No