missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium 4-(ethylthio)phenyltrifluoroborate, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5267703

 View more versions of this product

Catalog No. AAH5267703


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Potassium 4-(ethylthio)phenyltrifluoroborate
>300°C
MFCD04115771
potassium 4-ethylthiophenyl trifluoroborate, potassium 4-ethylthio phenyl trifluoroborate, potassium 4-ethylsulfanyl phenyl trifluoroboranuide, potassium 4-ethylthio phenyltrifluoroborate, potassium 4-ethylthio phenyl-trifluoroboranuide, potassium ion 4-ethylsulfanyl phenyl trifluoroboranuide, potassium 4-ethylsulfanylphenyl-tris fluoranyl boranuide, potassium 4-ethylsulfanyl phenyl trifluoro borate 1-
[B-](C1=CC=C(C=C1)SCC)(F)(F)F.[K+]
244.124
244.12
850623-75-5
C8H9BF3KS
1 g
BVLNLNRSQDZWIL-UHFFFAOYSA-N
potassium;(4-ethylsulfanylphenyl)-trifluoroboranuide
44717235
96%

Chemical Identifiers

850623-75-5
244.124
BVLNLNRSQDZWIL-UHFFFAOYSA-N
44717235
[B-](C1=CC=C(C=C1)SCC)(F)(F)F.[K+]
C8H9BF3KS
MFCD04115771
potassium 4-ethylthiophenyl trifluoroborate, potassium 4-ethylthio phenyl trifluoroborate, potassium 4-ethylsulfanyl phenyl trifluoroboranuide, potassium 4-ethylthio phenyltrifluoroborate, potassium 4-ethylthio phenyl-trifluoroboranuide, potassium ion 4-ethylsulfanyl phenyl trifluoroboranuide, potassium 4-ethylsulfanylphenyl-tris fluoranyl boranuide, potassium 4-ethylsulfanyl phenyl trifluoro borate 1-
potassium;(4-ethylsulfanylphenyl)-trifluoroboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions