missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium 4-fluorophenyltrifluoroborate, 95%, Thermo Scientific Chemicals
$109.10 - $238.52
Chemical Identifiers
CAS | 192863-35-7 |
---|---|
Molecular Formula | C6H4BF4K |
Molecular Weight (g/mol) | 202 |
MDL Number | MFCD01318170 |
InChI Key | PXKNUQDMYBUYSZ-UHFFFAOYSA-N |
Synonym | potassium 4-fluorophenyltrifluoroborate, potassium trifluoro 4-fluorophenyl borate, potassium trifluoro 4-fluorophenyl boranuide, potassium 4-fluorophenyl trifluoroborate, 4-fluorophenyl-trifluoroborate potassium salt, 4-fluorophenyltrifluoroboric acid potassium salt, 4-fluorobenzenetrifluoroboric acid potassium salt, pubchem11422, potassiotrifluoro 4-fluorophenyl boron v, potassium trifluoro-4-fluorophenyl boranuide |
PubChem CID | 23677644 |
IUPAC Name | potassium;trifluoro-(4-fluorophenyl)boranuide |
SMILES | [B-](C1=CC=C(C=C1)F)(F)(F)F.[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1765503
|
Thermo Scientific Chemicals
L1765503 |
1 g |
N/A
|
|
|||||
AAL1765506
|
Thermo Scientific Chemicals
L1765506 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
192863-35-7 | |
202 | |
PXKNUQDMYBUYSZ-UHFFFAOYSA-N | |
23677644 | |
[B-](C1=CC=C(C=C1)F)(F)(F)F.[K+] |
C6H4BF4K | |
MFCD01318170 | |
potassium 4-fluorophenyltrifluoroborate, potassium trifluoro 4-fluorophenyl borate, potassium trifluoro 4-fluorophenyl boranuide, potassium 4-fluorophenyl trifluoroborate, 4-fluorophenyl-trifluoroborate potassium salt, 4-fluorophenyltrifluoroboric acid potassium salt, 4-fluorobenzenetrifluoroboric acid potassium salt, pubchem11422, potassiotrifluoro 4-fluorophenyl boron v, potassium trifluoro-4-fluorophenyl boranuide | |
potassium;trifluoro-(4-fluorophenyl)boranuide |
Specifications
192863-35-7 | |
C6H4BF4K | |
1 g | |
potassium 4-fluorophenyltrifluoroborate, potassium trifluoro 4-fluorophenyl borate, potassium trifluoro 4-fluorophenyl boranuide, potassium 4-fluorophenyl trifluoroborate, 4-fluorophenyl-trifluoroborate potassium salt, 4-fluorophenyltrifluoroboric acid potassium salt, 4-fluorobenzenetrifluoroboric acid potassium salt, pubchem11422, potassiotrifluoro 4-fluorophenyl boron v, potassium trifluoro-4-fluorophenyl boranuide | |
[B-](C1=CC=C(C=C1)F)(F)(F)F.[K+] | |
202 | |
202 | |
Potassium 4-fluorophenyltrifluoroborate |
290°C to 292°C | |
MFCD01318170 | |
7783544 | |
PXKNUQDMYBUYSZ-UHFFFAOYSA-N | |
potassium;trifluoro-(4-fluorophenyl)boranuide | |
23677644 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only