missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium (4-Methoxyphenyl)trifluoroborate 98.0+%, TCI America™
Supplier: TCI America P18201G
Specifications
| Potassium (4-Methoxyphenyl)trifluoroborate | |
| 192863-36-8 | |
| 1 g | |
| MFCD04112717 | |
| XNYMCUFKZHRYID-UHFFFAOYSA-N | |
| potassium;trifluoro-(4-methoxyphenyl)boranuide | |
| 23683762 | |
| ≥98.0% (HPLC,T) |
| White | |
| Crystalline Powder | |
| C7H7BF3KO | |
| potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide | |
| [B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+] | |
| 214.036 | |
| 214.04 |
Chemical Identifiers
| 192863-36-8 | |
| 214.036 | |
| XNYMCUFKZHRYID-UHFFFAOYSA-N | |
| 23683762 | |
| [B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+] |
| C7H7BF3KO | |
| MFCD04112717 | |
| potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide | |
| potassium;trifluoro-(4-methoxyphenyl)boranuide |
Safety and Handling
TSCA : No