missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium 4-methoxyphenyltrifluoroborate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2738406

 View more versions of this product

Catalog No. AAH2738406


Explore available promotions

Description

Description

It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Potassium 4-methoxyphenyltrifluoroborate
>300°C
MFCD04112717
potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide
[B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+]
214.036
214.04
192863-36-8
C7H7BF3KO
5 g
XNYMCUFKZHRYID-UHFFFAOYSA-N
potassium;trifluoro-(4-methoxyphenyl)boranuide
23683762
95%

Chemical Identifiers

192863-36-8
214.036
XNYMCUFKZHRYID-UHFFFAOYSA-N
23683762
[B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+]
C7H7BF3KO
MFCD04112717
potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide
potassium;trifluoro-(4-methoxyphenyl)boranuide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only