missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 223861000

 View more versions of this product

Catalog No. AC223861000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7440-09-7
K
MFCD00133776
01,905; 06,274; 12,400; 13,170; 14,308; 15,269
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
[K]
39.10
CHEBI:26216
Plastic bottle
Potassium
98%
K
100 g
15, 7725
ZLMJMSJWJFRBEC-UHFFFAOYSA-N
potassium
5462222
39.1
Chunk

Chemical Identifiers

7440-09-7
39.10
ZLMJMSJWJFRBEC-UHFFFAOYSA-N
5462222
potassium
K
MFCD00133776
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
CHEBI:26216
[K]
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only