missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Antimony Tartrate Trihydrate, ACS, 99-103%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A1561125GM

 View more versions of this product

Catalog No. 18612451

Explore available promotions

Description

Description

Specifications

Specifications

28300-74-5
0.027
125 g
ACS
1
Poly Bottle
99 to 103%

Chemical Identifiers

28300-74-5
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.